Learn More
Sino Biological E. coli acnB Matched ELISA Antibody Pair Set 5 Plates
Supplier:  Sino Biological SEK131025
Encompass_Preferred
The Escherichia coli acnB ELISA Pair Set is for the quantitative determination of Escherichia coli acnB.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Escherichia coli acnB coated on a 96-well plate. Standards and samples are added to the wells, and any Escherichia coli acnB present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Escherichia coli acnB monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Escherichia coli acnB present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86706