Learn More
New England Biolabs, Inc. E.coli DNA Ligase – 200 units
SDP
Catalog No. 50811612
Encompass
E. coli DNA Ligase catalyzes the formation of a phosphodiester bond between the 5'-phosphate and the 3'-hydroxyl of two adjacent DNA strands in duplex DNA with cohesive ends. It is not appreciably active on blunt-ended substrates. E. coli DNA Ligase uses NAD as a cofactor and can be heat-inactivated.
Catalog No. 50-811-612 Supplier New England Biolabs, Inc. Supplier No. M0205S
E. coli DNA Ligase catalyzes the formation of a phosphodiester bond between the 5'-phosphate and the 3'-hydroxyl of two adjacent DNA strands in duplex DNA with cohesive ends. It is not appreciably active on blunt-ended substrates. E. coli DNA Ligase uses NAD as a cofactor and can be heat-inactivated.