Learn More
Qiagen Beverly Inc E. COLI DNA LIGASE
Catalog No. 50305902
Encompass
DNA Ligase; E. Coli DNA Ligase; Conc. 10,000 U/ul; 2,500U; incl. 10X buffer
Catalog No. 50-305-902 Supplier Qiagen Beverly Inc Supplier No. L6090L
DNA Ligase; E. Coli DNA Ligase; Conc. 10,000 U/ul; 2,500U; incl. 10X buffer