Learn More
Bioworld E.COLI DNAJ (HSP40) (RECOM
Supplier:  Bioworld 220600561
Encompass_Preferred
E.COLI DNAJ (HSP40), (RECOMBINANT): Recombinant Dna J Source: E.Coli Description : DnaJ, Heat shock protein, functions in association with DnaK(Hsp70) molecular chaperone to facilitate protein folding. p70 chaperone. DnaJ plays a key...
Catalog No. 50-253-153