Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E.coli Genomic DNA

Catalog No. AAJ14380MA Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ14380MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ14380MA Supplier Thermo Scientific Supplier No. J14380MA
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 9007-49-2 | C15H31N3O13P2

 

  • Single chromosome
  • Genome length: 4600000 bp
  • Fragmented to some degree during purification yet it is characterized as high molecular weight DNA
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9007-49-2

Specifications

Product Type Genomic DNA
Quantity 10 mg
Form Powder

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.