Learn More
Bioworld E.COLI GROES (HSP10) (RECO
Supplier:  Bioworld 220600861
Encompass_Preferred
E.COLI GROES (HSP10), (RECOMBINANT): Recombinant GroES Source: E.Coli Description : GroES protein is the co-chaperonin of GroES in E.coli and assists protein folding. GroEL mediated folding requires the co-chaperonin GroES which is e...
Catalog No. 50-253-187