Learn More
Bon Opus Biosciences Recombinant E.coli GUSB/Beta-Glucuronidase(N-His) 1mg
SDP
Supplier:  Bon Opus Biosciences CH851MG
Encompass_Preferred
Recombinant E.coli beta-Glucuronidase is produced by our E.coli expression system and the target gene encoding Met1-Gln603 is expressed with a 6His tag at the N-terminus. Accession# P05804. Formulation: Supplied as a 0.2 um filtered solution of PBS, pH7.4 Bon Opus Cat# CH85
Catalog No. 50-217-2796
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.