Learn More
Abcam E. coli HCP ELISA Kit (host cell protein), 1 x 96 Tests
Supplier:  Abcam AB2409971X96TESTS
Encompass_Preferred
E. coli HCP ELISA Kit (host cell protein) ab240997 is a sandwich ELISA used to quantify host cell protein (HCP) contamination in bulk products expressed in E.coli expression systems with a sensitivity of 0.32 ng/mL. - Wide dynamic range – quantifies 3–810 ng/mL - Colorimetric assay - 450 nm readout - works on any plate reader - Recommended to use with research bioproducts including in-process samples, downstream samples and spike recovery testing
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2239517