Learn More
Enzo Life Sciences E. coli host cell proteins polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2620100
Encompass_Preferred
Immunogen: E. coli host cell proteins. Species reactivity: E. coli. Specificity: Recognizes E. coli host cell proteins. Applications: WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 10ppm Kathon.
Catalog No. 50-201-2807