Learn More
Enzo Life Sciences E. coli host cell proteins polyclonal antibody (biotin conjugate) (100µg)
SDP
Catalog No. 502012808
Encompass_Preferred
Immunogen: E. coli host cell proteins. Species reactivity: E. coli. Specificity: Recognizes E. coli host cell proteins. Applications: WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBST with 1% BSA containing 10ppm Kathon.
Catalog No. 50-201-2808 Supplier Enzo Life Sciences Supplier No. ENZABS2630100
Immunogen: E. coli host cell proteins. Species reactivity: E. coli. Specificity: Recognizes E. coli host cell proteins. Applications: WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBST with 1% BSA containing 10ppm Kathon.