Learn More
Enzo Life Sciences E. coli host cell protein standards (25µg)
SDP
Catalog No. 502013914
Encompass_Preferred
E. coli host cell protein standards are native proteins that can be used as an internal control for the Western blot analysis of CHO Host Cell Proteins or as standards in an ELISA. E. coli host cell protein standards are native proteins that can be used as an internal control for the Western blot analysis of CHO Host Cell Proteins or as standards in an ELISA. Formulation: Liquid. In PBS containing 10 ppm Kathon. Source: Purified from E. coli host cells. Long Term Storage: -20°C. Applications: ELISA, WB .
Catalog No. 50-201-3914 Supplier Enzo Life Sciences Supplier No. ENZPRT1210025
E. coli host cell protein standards are native proteins that can be used as an internal control for the Western blot analysis of CHO Host Cell Proteins or as standards in an ELISA. E. coli host cell protein standards are native proteins that can be used as an internal control for the Western blot analysis of CHO Host Cell Proteins or as standards in an ELISA. Formulation: Liquid. In PBS containing 10 ppm Kathon. Source: Purified from E. coli host cells. Long Term Storage: -20°C. Applications: ELISA, WB .