Learn More
Enzo Life Sciences E. coli host cell protein ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ENZKIT1270001
Encompass_Preferred
Sensitive (30 ng/ml) ELISA for  the quantitative determination of host cell protein contamination in bulk products expressed in E. coli expression systems. The E. coli host cell protein ELISA kit is a complete, colorimetric, immunometric immunoassay kit for the quantitative determination of E. coli host cell protein contamination with results in 3 hours. Sensitivity: 30 ng/ml (range 3-810 ng/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Contents: 1 x 96 well plate, protein standards, 5x dilution buffer, 10x PBS-T, reporting antibody, streptavidin-HRP conjugate, TMB substrate, stop solution, plate sealer
Catalog No. NC1673935