Learn More
Sino Biological Anti-LON PROTEASE Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 40219RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Escherichia coli LON PROTEASE (Catalog#40219-E08E; P0A9M0; Met1-Lys784). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3735