Learn More
Invivogen Standard LPS, E. coli K12, 5 mg
SDP
Supplier:  Invivogen TLRLEKLPS
Encompass
LPS-EK is a standard lipopolysaccharide (LPS) preparation extracted by a phenol-water mixture. LPS-EK contains other bacterial components, such as lipoproteins, and therefore stimulates both TLR4 and TLR2.
Catalog No. NC9355258
 Restricted Item