Learn More
Biotang Inc E. Coli Lysis Buffer (Tris-HCl 50 mM,1% Triton,EDTA 1 mM, Beta-mercaptoethanol 10 mM), 250 ml
Catalog No. 50155919 Shop All BIOTANG Inc Products
Encompass_Preferred
E. Coli Lysis Buffer (Tris-HCl 50 mM,1% Triton,EDTA 1 mM, Beta-mercaptoethanol 10 mM), 250 ml.
Catalog No. 50-155-919 Supplier Biotang Inc Supplier No. BTBB639
E. Coli Lysis Buffer (Tris-HCl 50 mM,1% Triton,EDTA 1 mM, Beta-mercaptoethanol 10 mM), 250 ml.