Learn More
Biotang Inc E. Coli Lysis Buffer-II (Phosphate buffer pH 7.4 10 mM, KCL 2.7 mM,NaCl 140 mM, 1% Triton , 250 ml
Catalog No. 50155920 Shop All BIOTANG Inc Products
Encompass_Preferred
E. Coli Lysis Buffer-II (Phosphate buffer pH 7.4 10 mM, KCL 2.7 mM,NaCl 140 mM, 1% Triton , 250 ml.
Catalog No. 50-155-920 Supplier Biotang Inc Supplier No. BTBB640
E. Coli Lysis Buffer-II (Phosphate buffer pH 7.4 10 mM, KCL 2.7 mM,NaCl 140 mM, 1% Triton , 250 ml.