Learn More
Bioworld E. coli Lysis Buffer, ii, pH 7.4, 250 mL
Catalog No. 50253023
Encompass_Preferred
E. coli Lysis Buffer II Sample buffer for protein extraction and purification of GST-fusion protein. Technical Information:Phosphate Buffer, pH 7.4 (10mM)Triton X 100 (1%)KCl (2.7mM)NaCl (140mM)
Catalog No. 50-253-023 Supplier Bioworld Supplier No. 220400281
E. coli Lysis Buffer II Sample buffer for protein extraction and purification of GST-fusion protein. Technical Information:Phosphate Buffer, pH 7.4 (10mM)Triton X 100 (1%)KCl (2.7mM)NaCl (140mM)