Learn More
Bioworld E. coli Lysis Buffer, ii, pH 8.0, 250 mL
Catalog No. 50253025
Encompass_Preferred
E. coli Lysis Buffer II Sample buffer for protein extraction and purification of GST-fusion protein. Technical Information:Tris-HCl, pH 8.0 (50mM)Triton X 100 (1%)EDTA, pH 8.0 (1mM)Beta-mercaptoethanol (10mM)
Catalog No. 50-253-025 Supplier Bioworld Supplier No. 220400291
E. coli Lysis Buffer II Sample buffer for protein extraction and purification of GST-fusion protein. Technical Information:Tris-HCl, pH 8.0 (50mM)Triton X 100 (1%)EDTA, pH 8.0 (1mM)Beta-mercaptoethanol (10mM)