Learn More
New England Biolabs, Inc. NEBExpress E. coli Lysis Reagent 500 mL (5 x 100 mL)
SDP
Catalog No. 502172599
Encompass
NEBExpress E. coli Lysis Reagent is a chemical lysis solution composed of a proprietary mix of non-ionic and zwitterionic detergents and Tris-based buffer. It allows disruption of E. coli cells without denaturing soluble proteins.
Catalog No. 50-217-2599 Supplier New England Biolabs, Inc. Supplier No. P8116L
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
NEBExpress E. coli Lysis Reagent is a chemical lysis solution composed of a proprietary mix of non-ionic and zwitterionic detergents and Tris-based buffer. It allows disruption of E. coli cells without denaturing soluble proteins.