Learn More
Sino Biological E. coli Maltose Binding Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 69088E07E1
Encompass_Preferred
A DNA sequence encoding the E. coli malE (WP_052916395.1) (Glu32-Lys399) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-255-2907