Learn More
Seracare Life Sciences Inc E COLI O111:H8 SPECIES POS CON
Catalog No. 5067466
Encompass_Preferred
Heat-killed Escherichia coli O111:H8 cells in dextran solution. It is intended for use as a positive control in immunoassays designed for E.coli O111 detection.
This item is not returnable. View return policy
Catalog No. 50-674-66 Supplier Seracare Life Sciences Inc Supplier No. 53700014
Heat-killed Escherichia coli O111:H8 cells in dextran solution. It is intended for use as a positive control in immunoassays designed for E.coli O111 detection.