Learn More
Seracare Life Sciences Inc E COLI O157:H7 POS CONTROL
Supplier:  Seracare Life Sciences Inc 53700013
Encompass_Preferred
Heat-killed Escherichia coli O157:H7 cells. It consists ofat least 3.0 x 109 cfu/mL(1 mL of approximately 2% w/v wet packed cells) in dextran solution. It is intended for use as a positive control in immunoassays designed for E.coli 0157 detection.
Catalog No. 50-674-65