Learn More
Seracare Life Sciences Inc E COLI O26:H11 SPECIES POS CON
Catalog No. 5067467
Encompass_Preferred
Heat-killed Escherichia coli O26:H11 cells in dextran solution. It is intended for use as a positive control in immunoassays designed for E.coli O26 detection.
This item is not returnable. View return policy
Catalog No. 50-674-67 Supplier Seracare Life Sciences Inc Supplier No. 53700015
Heat-killed Escherichia coli O26:H11 cells in dextran solution. It is intended for use as a positive control in immunoassays designed for E.coli O26 detection.