Learn More
New England Biolabs, Inc. E.coli RNA Polymerase, Core Enzyme – 100 units
SDP
Catalog No. 50591123
Encompass
E. coli RNA Polymerase, Core Enzyme consists of 5 subunits designated α, α, βʹ , β, and ω.
Catalog No. 50-591-123 Supplier New England Biolabs, Inc. Supplier No. M0550S
E. coli RNA Polymerase, Core Enzyme consists of 5 subunits designated α, α, βʹ , β, and ω.