Learn More
New England Biolabs, Inc. E.coli RNA Polymerase, Holoenzyme – 50 units
SDP
Catalog No. 50591124
Encompass
E. coli RNA Polymerase, Holoenzyme is the core enzyme saturated with sigma factor 70. The Holoenzyme initiates RNA synthesis from sigma 70 specific bacterial and phage promoters. E. coli RNA Polymerase, Core Enzyme consists of 5 subunits designated α, α, βʹ , β, and ω. The enzyme is free of sigma factor and does not recognize any specific bacterial or phage DNA promoters. The enzyme retains the ability to transcribe RNA from nonspecific initiation sequences. Addition of sigma factors will allow the enzyme to initiate RNA synthesis from specific bacterial and phage promoters. The core enzyme has a molecular weight of approximately 400 kDa.
Catalog No. 50-591-124 Supplier New England Biolabs, Inc. Supplier No. M0551S
E. coli RNA Polymerase, Holoenzyme is the core enzyme saturated with sigma factor 70. The Holoenzyme initiates RNA synthesis from sigma 70 specific bacterial and phage promoters. E. coli RNA Polymerase, Core Enzyme consists of 5 subunits designated α, α, βʹ , β, and ω. The enzyme is free of sigma factor and does not recognize any specific bacterial or phage DNA promoters. The enzyme retains the ability to transcribe RNA from nonspecific initiation sequences. Addition of sigma factors will allow the enzyme to initiate RNA synthesis from specific bacterial and phage promoters. The core enzyme has a molecular weight of approximately 400 kDa.