Learn More
Alpco Ltd E. coli Stool Panel I PCR Kit
SDP
Catalog No. NC2284600
Encompass
The E. coli Stool Panel I PCR Kit is a multiplex real-time PCR assay for the direct, qualitative detection and differentiation of genes encoding the virulence-factors of EPEC and STEC, EPEC in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures.
This item is not returnable. View return policy
Catalog No. NC2284600 Supplier Alpco Ltd Supplier No. 86ECPHU100
The E. coli Stool Panel I PCR Kit is a multiplex real-time PCR assay for the direct, qualitative detection and differentiation of genes encoding the virulence-factors of EPEC and STEC, EPEC in human stool samples. Research Use Only. Not for Use in Diagnostic Procedures.