Learn More
Sino Biological Anti-Shiga toxin II subunit B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 40019T1620
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant recombinant Enterohemorrhagic E. coli (EHEC) stx2B (Catalog# 40019-E08E; Q93EY4; Met1-Asp89). stx2B specific IgG was purified by Enterohemorrhagic E. coli (EHEC) stx2B affinity chromatography.
Catalog No. 50-255-3752