Learn More
New England Biolabs, Inc. E. coli tRNA - 1 mg
SDP
Catalog No. 5000460441
Encompass
E. coli tRNA is a mix of tRNAs isolated from E. coli and purified by chromatography to remove contaminating proteins, including nucleases, as well as other nucleic acids. This tRNA formulation sustains translation in E. coli-based in vitro protein synthesis reactions (also referred to as cell-free protein synthesis or CFPS). It is also suited for molecular biology applications that require highly pure and active prokaryotic tRNA.
  • Highly active tRNA for in vitro translation
  • Purified by chromatography, no organic solvents
  • Highly pure with undetectable levels of nucleases and proteases
  • Manufactured with reduced environmental impact
  • Compatible with a variety of molecular biology workflows
Catalog No. 50-004-60441 Supplier New England Biolabs, Inc. Supplier No. N6845S
E. coli tRNA is a mix of tRNAs isolated from E. coli and purified by chromatography to remove contaminating proteins, including nucleases, as well as other nucleic acids. This tRNA formulation sustains translation in E. coli-based in vitro protein synthesis reactions (also referred to as cell-free protein synthesis or CFPS). It is also suited for molecular biology applications that require highly pure and active prokaryotic tRNA.
  • Highly active tRNA for in vitro translation
  • Purified by chromatography, no organic solvents
  • Highly pure with undetectable levels of nucleases and proteases
  • Manufactured with reduced environmental impact
  • Compatible with a variety of molecular biology workflows