Learn More
E. coli in Water, PT- Quantitative WP Proficiency Testing Material, MilliporeSigma™ Supelco™
Catalog No. 111004543 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2/Pk.
Catalog No. Quantity
11-100-4543 2/Pk.
1 options
Catalog No. 11-100-4543 Supplier MilliporeSigma Supelco Supplier No. MIC0032EA
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
Set contains 2 sterile screw cap vials each containing a Vitroid™ (a small disk containing the possible organisms) and 2 - 99 mL bottles of phosphate buffer.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2/Pk.
Concentration or Composition (by Analyte or Components) Total coliforms 20.0 - 2400 CFU/100 mL; Total Coliform, MPN 20.0 - 2400 MPN/100 mL; Escherichia coli 20.0 - 2400 CFU/100 mL; Escherichia coli, MPN 20.0 - 2400 MPN/100 mL; Fecal coliforms 20.0 - 2400 CFU/100 mL; Fecal coliform, MPN 20.0 - 2400 MPN/100 mL
Vitroid is a trademark of Sigma-Aldrich Co. LLC