Learn More
Med Vet International Jorvet Flexy E-Collar, Small, J1095B, Each
Catalog No. 5000495709
Encompass_Preferred
The Jorvet small flexible e-collar is made of a non-woven water resistant material. It is lightly padded for comfort and better protection. The shoelace drawstring wroks especially well to secure the e-collar on cats.
Catalog No. 50-004-95709 Supplier Med Vet International Supplier No. JOR1095B
The Jorvet small flexible e-collar is made of a non-woven water resistant material. It is lightly padded for comfort and better protection. The shoelace drawstring wroks especially well to secure the e-collar on cats.