Learn More
Medchemexpress LLC (E)-Cyclooct-2-en-1-yl (2-(2-(2-(2-aminoethoxy)ethoxy)ethoxy)ethyl)carbamate | 98.4% | 344.45 | C17H32N2O5 | 10 MG
SDP
Catalog No. 5000288206
Encompass_Preferred
TCO-PEG3-NH2 is a click-chemistry reagent composed of a trans-cyclooctene (TCO) group linked to a three-unit polyethylene glycol spacer terminating in a primary amine. It enables fast bioorthogonal inverse-electron-demand Diels-Alder (iEDDA) reactions with tetrazine-functionalized partners and amine-mediated conjugation for site-specific labeling of biomolecules.
  • Enables rapid iEDDA bioorthogonal chemistry for fast conjugation.
  • Terminal primary amine allows amide bond formation with carboxyl groups.
  • Three-unit PEG spacer improves aqueous solubility and reduces steric hindrance.
  • Suitable for site-specific labeling and bioconjugation workflows.
  • Typically supplied as a small solid quantity for laboratory use.
Catalog No. 50-002-88206 Supplier Medchemexpress LLC Supplier No. HY157006A10MG
TCO-PEG3-NH2 is a click-chemistry reagent composed of a trans-cyclooctene (TCO) group linked to a three-unit polyethylene glycol spacer terminating in a primary amine. It enables fast bioorthogonal inverse-electron-demand Diels-Alder (iEDDA) reactions with tetrazine-functionalized partners and amine-mediated conjugation for site-specific labeling of biomolecules.
  • Enables rapid iEDDA bioorthogonal chemistry for fast conjugation.
  • Terminal primary amine allows amide bond formation with carboxyl groups.
  • Three-unit PEG spacer improves aqueous solubility and reduces steric hindrance.
  • Suitable for site-specific labeling and bioconjugation workflows.
  • Typically supplied as a small solid quantity for laboratory use.