Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (E)-Cyclooct-4-Enol

Catalog No. 76439650MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-439-650MG 10 mg
76-439-610MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 76-439-650MG Supplier Sigma Aldrich Supplier No. 76439650MG
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hydroxyl modified cyclooctene derivative. Cyclooctenes are useful in strain-promoted copper-free click chemistry cycloaddition reactions with 1,2,4,5-tetrazines. This cyclooctene will react with tetrazine functionalized compounds or biomolecules without the need for a catalyst to result in a stable covalent linkage. The 4+2 inverse electron demand Diels-Alder cycloaddition between trans-cyclooctene and tetrazines is the fastest biologically compatible ligation technology reported and has had many applications in biological labeling and imaging.

Specifications

CAS 1191901-33-3
Molecular Formula C8H14O
Linear Formula C8H14O
Quantity 10 mg
Synonym (4E)-4-Cycloocten-1-ol; trans-Cyclooctenol; TCO-OH; rel-(1R,4E,pR)-Cyclooct-4-enol
Molecular Weight (g/mol) 126.2 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.