Learn More
Medchemexpress LLC HY-50876 10mg Medchemexpress, (E)-Daporinad CAS:658084-64-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5087610MG
Encompass_Preferred
Medchemexpress, HY-50876 10mg (E)-Daporinad CAS:658084-64-1 (E)-Daporinad (FK866) is an effective inhibitor of nicotinamide phosphoribosyltransferase (NMPRTase; Nampt) with an IC50 of 0.09 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9345
 Restricted Item