Learn More
Biolegend PE/Dazzle™ 594 anti-mouse CD279 (PD-1)
Supplier:  Biolegend 135228
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD279 (PD-1) [29F.1A12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC0990770