Learn More
Medchemexpress LLC (E,E)-4,8,12-Trimethyl-1,3,7,11-tridecatetraene | 62235-06-7 | 218.38 | 100 MG
SDP
Catalog No. 5000364971
Encompass_Preferred
(E,E)-4,8,12-Trimethyl-1,3,7,11-tridecatetraene (TMTT) is a sesquiterpene and a major herbivore-induced plant volatile. It functions in plant defense by directly repelling pests and indirectly by attracting natural enemies of insects. This compound has been shown to attract Asian corn borer (ACB, Ostrinia furnacalis) larvae, and external supplementation can impede their ability to locate pre-infested maize. Its target is insecticide, with a pathway classified as "Others".
  • Major herbivore-induced plant volatile
  • Assists in plant defense
  • Acts as a repellant
  • Recruits natural enemies of insects
  • Attracts Asian corn borer larvae
  • Hinders larvae's ability to locate pre-infested maize
  • Targeted as an insecticide
Catalog No. 50-003-64971 Supplier Medchemexpress LLC Supplier No. 100MGHYW585959
(E,E)-4,8,12-Trimethyl-1,3,7,11-tridecatetraene (TMTT) is a sesquiterpene and a major herbivore-induced plant volatile. It functions in plant defense by directly repelling pests and indirectly by attracting natural enemies of insects. This compound has been shown to attract Asian corn borer (ACB, Ostrinia furnacalis) larvae, and external supplementation can impede their ability to locate pre-infested maize. Its target is insecticide, with a pathway classified as "Others".
  • Major herbivore-induced plant volatile
  • Assists in plant defense
  • Acts as a repellant
  • Recruits natural enemies of insects
  • Attracts Asian corn borer larvae
  • Hinders larvae's ability to locate pre-infested maize
  • Targeted as an insecticide