Learn More
Medchemexpress LLC (E,E)-4,8,12-Trimethyl-1,3,7,11-tridecatetraene | 62235-06-7 | 98% | 218.38 | 25 MG
SDP
Catalog No. 5000366108
Encompass_Preferred
(E,E)-4,8,12-Trimethyl-1,3,7,11-tridecatetraene (TMTT) is a sesquiterpene and a major herbivore-induced plant volatile. It functions in plant defense by acting as a repellent and by recruiting the natural enemies of insects. TMTT has been shown to attract Asian corn borer (ACB, Ostrinia furnacalis) larvae, and its external supplementation can hinder the larvae's ability to locate pre-infested maize.
  • Functions as a sesquiterpene
  • Major herbivore-induced plant volatile
  • Aids in plant defense as a repellent
  • Recruits natural enemies of insects
  • Attracts Asian corn borer larvae
  • Hindered larvae's ability to find pre-infested maize upon supplementation
Catalog No. 50-003-66108 Supplier Medchemexpress LLC Supplier No. 25MGHYW585959
(E,E)-4,8,12-Trimethyl-1,3,7,11-tridecatetraene (TMTT) is a sesquiterpene and a major herbivore-induced plant volatile. It functions in plant defense by acting as a repellent and by recruiting the natural enemies of insects. TMTT has been shown to attract Asian corn borer (ACB, Ostrinia furnacalis) larvae, and its external supplementation can hinder the larvae's ability to locate pre-infested maize.
  • Functions as a sesquiterpene
  • Major herbivore-induced plant volatile
  • Aids in plant defense as a repellent
  • Recruits natural enemies of insects
  • Attracts Asian corn borer larvae
  • Hindered larvae's ability to find pre-infested maize upon supplementation