Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (E,E)-8,10-dodecadien-1-ol | 33956-49-9 | 95.2% | 182.30 g·mol⁻¹ | C12H22O | 25G
SDP

Catalog No. 5000236855
Encompass_Preferred

Codlemone (Codlelure) is (E,E)-8,10-dodecadien-1-ol (CAS 33956-49-9), the primary sex pheromone of the codling moth (Cydia pomonella). Supplied as a research-grade analytical standard for pheromone research, behavioral studies, and formulation testing.

  • High-purity analytical standard (95.2%).
  • Chemical formula C12H22O and molecular weight 182.30 g·mol⁻¹.
  • Suitable for pheromone-based behavioral assays and field trials.
  • Packaged as 25 G for laboratory use.
  • Physical state: solid below 30°C, liquid above 32°C.
  • CAS registry number 33956-49-9 for unambiguous identification.

Catalog No. 50-002-36855 Supplier Medchemexpress LLC Supplier No. HY12446625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Codlemone (Codlelure) is (E,E)-8,10-dodecadien-1-ol (CAS 33956-49-9), the primary sex pheromone of the codling moth (Cydia pomonella). Supplied as a research-grade analytical standard for pheromone research, behavioral studies, and formulation testing.

  • High-purity analytical standard (95.2%).
  • Chemical formula C12H22O and molecular weight 182.30 g·mol⁻¹.
  • Suitable for pheromone-based behavioral assays and field trials.
  • Packaged as 25 G for laboratory use.
  • Physical state: solid below 30°C, liquid above 32°C.
  • CAS registry number 33956-49-9 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.