Learn More
Apexbio Technology LLC (E,E)-Bisdemethoxycurcumin 33171-05-0 50mg
SDP
Catalog No. 5000094945
Encompass_Preferred
(E E)-Bisdemethoxycurcumin (CAS 33171-05-0) is a naturally derived curcuminoid and a structural analog within the demethoxycurcumin class Research demonstrates that this compound modulates multiple inflammatory signaling pathways and alters the expression of various inflammatory mediators It has been shown to induce cell cycle arrest promote apoptosis in cancer cells and inhibit proliferation-related pathways (E E)-Bisdemethoxycurcumin is widely utilized in studies investigating the molecular mechanisms of inflammation tumor cell biology and the pharmacological properties of curcuminoid derivatives
Catalog No. 50-000-94945 Supplier Apexbio Technology LLC Supplier No. A465250
(E E)-Bisdemethoxycurcumin (CAS 33171-05-0) is a naturally derived curcuminoid and a structural analog within the demethoxycurcumin class Research demonstrates that this compound modulates multiple inflammatory signaling pathways and alters the expression of various inflammatory mediators It has been shown to induce cell cycle arrest promote apoptosis in cancer cells and inhibit proliferation-related pathways (E E)-Bisdemethoxycurcumin is widely utilized in studies investigating the molecular mechanisms of inflammation tumor cell biology and the pharmacological properties of curcuminoid derivatives