Learn More
Medchemexpress LLC (E,E)-Bisdemethoxycurcumin (Standard) | 33171-05-0 | 98.5% | 308.33 | 1MG
SDP
Catalog No. 5000431625
Encompass_Preferred
(E,E)-Bisdemethoxycurcumin (Standard) is the analytical standard of (E,E)-Bisdemethoxycurcumin, designed for research and analytical applications. This curcumin derivative possesses anti-inflammatory and anticancer activities. It serves as a reference standard for assays and is commonly used in qualitative, quantitative, and methodological research experiments like HPLC, GC, and MS.
  • Intended for research and analytical applications
  • Possesses anti-inflammatory and anticancer activities
  • Analytical standard grade for assays
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-004-31625 Supplier Medchemexpress LLC Supplier No. HYN0007S1MG
(E,E)-Bisdemethoxycurcumin (Standard) is the analytical standard of (E,E)-Bisdemethoxycurcumin, designed for research and analytical applications. This curcumin derivative possesses anti-inflammatory and anticancer activities. It serves as a reference standard for assays and is commonly used in qualitative, quantitative, and methodological research experiments like HPLC, GC, and MS.
  • Intended for research and analytical applications
  • Possesses anti-inflammatory and anticancer activities
  • Analytical standard grade for assays
  • Suitable for HPLC, GC, and MS experiments