Learn More
Echelon Biosciences Research Labs (E,E) FARNESOIC ACID 100 mg
SDP
Supplier:  Echelon Biosciences Research Labs S0151100MG
Encompass
Farnesoic acid ((2E,6E)-3,7,11-trimethyldodeca-2,6,10-trienoic acid) is the substrate of farnesoic acid O-methyltransferase which produces the crustacean reproductive hormone, methyl farnesoate (MF). MF is responsible for enhancing reproductive maturation, maintaining juvenile morphology, and influencing male sex determination. Farnesoic acid also inhibits filiment formation in C. albicans. This item is non returnable
Catalog No. 50-720-749