Learn More
Medchemexpress LLC (E, E)-Farnesyl bromide | 28290-41-7 | MFCD00054465 | 95% | 285.26 | 100 MG
SDP
Catalog No. 5000366831
Encompass_Preferred
Trans,trans-Farnesyl bromide was used in the preparation of S-trans, trans-farnesyl-L-cysteine methylester, a post translational modified amino acid. It is also a starting reagent for the synthesis of (±)-farnesiferol A and (±)-farnesiferol C prenylated peptides.
  • Preparation of S-trans, trans-farnesyl-L-cysteine methylester
  • Starting reagent for the synthesis of (±)-farnesiferol A and (±)-farnesiferol C prenylated peptides
Catalog No. 50-003-66831 Supplier Medchemexpress LLC Supplier No. HYW008629100MG
Trans,trans-Farnesyl bromide was used in the preparation of S-trans, trans-farnesyl-L-cysteine methylester, a post translational modified amino acid. It is also a starting reagent for the synthesis of (±)-farnesiferol A and (±)-farnesiferol C prenylated peptides.
  • Preparation of S-trans, trans-farnesyl-L-cysteine methylester
  • Starting reagent for the synthesis of (±)-farnesiferol A and (±)-farnesiferol C prenylated peptides