Learn More
Medchemexpress LLC (E, E)-Farnesyl bromide | 28290-41-7 | MFCD00054465 | 95% | 285.26 | 50 MG
SDP
Catalog No. 5000451604
Encompass_Preferred
Farnesyl bromide is a chemical compound used in various laboratory applications. It is identified by its CAS number 28290-41-7 and has a molecular weight of 285.26. This substance is classified as causing skin irritation and serious eye damage, and it may also cause respiratory irritation. It demonstrates chemical stability under recommended storage conditions.
  • Causes skin irritation
  • Causes serious eye damage
  • May cause respiratory irritation
  • Stable under recommended storage conditions
  • Used in the preparation of S-trans, trans-farnesyl-L-cysteine methylester
  • Used in the preparation of umbelliprenin
  • Starting reagent for the synthesis of (±)-farnesiferol A and (±)-farnesiferol C prenylated peptides
Catalog No. 50-004-51604 Supplier Medchemexpress LLC Supplier No. HYW00862950MG
Farnesyl bromide is a chemical compound used in various laboratory applications. It is identified by its CAS number 28290-41-7 and has a molecular weight of 285.26. This substance is classified as causing skin irritation and serious eye damage, and it may also cause respiratory irritation. It demonstrates chemical stability under recommended storage conditions.
  • Causes skin irritation
  • Causes serious eye damage
  • May cause respiratory irritation
  • Stable under recommended storage conditions
  • Used in the preparation of S-trans, trans-farnesyl-L-cysteine methylester
  • Used in the preparation of umbelliprenin
  • Starting reagent for the synthesis of (±)-farnesiferol A and (±)-farnesiferol C prenylated peptides