Learn More
Echelon Biosciences Research Labs (E,E) METHYL FARNESOATE 10 mg
SDP
Catalog No. 50720750
Encompass
Methyl farnesoate (MF) is a crustacean reproductive hormone that is structurally similar to insect juvenile hormone. It is responsible for enhancing reproductive maturation, maintaining juvenile morphology, and influencing male sex determination. Exposure of female Daphnids to increasing levels of MF increases the percentage of males in a brood in a dose-dependant manner. MF is endogenously produced in the mandibular organ and environmental factors such as salinity and temperature can influence hemolyph levels. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-750 Supplier Echelon Biosciences Research Labs Supplier No. S015310MG
Methyl farnesoate (MF) is a crustacean reproductive hormone that is structurally similar to insect juvenile hormone. It is responsible for enhancing reproductive maturation, maintaining juvenile morphology, and influencing male sex determination. Exposure of female Daphnids to increasing levels of MF increases the percentage of males in a brood in a dose-dependant manner. MF is endogenously produced in the mandibular organ and environmental factors such as salinity and temperature can influence hemolyph levels. This item is non returnable