Learn More
Echelon Biosciences Research Labs (E,E) METHYL FARNESOATE 100 mg
SDP
Supplier:  Echelon Biosciences Research Labs S0153100MG
Encompass
Methyl farnesoate (MF) is a crustacean reproductive hormone that is structurally similar to insect juvenile hormone. It is responsible for enhancing reproductive maturation, maintaining juvenile morphology, and influencing male sex determination. Exposure of female Daphnids to increasing levels of MF increases the percentage of males in a brood in a dose-dependant manner. MF is endogenously produced in the mandibular organ and environmental factors such as salinity and temperature can influence hemolyph levels. This item is non returnable
Catalog No. 50-720-751