Learn More
Medchemexpress LLC E/Envelope Dengue v 100ug
SDP
Supplier:  Medchemexpress LLC HYP74194100UG
Encompass_Preferred
E/Envelope Protein Dengue virus 2 (429a a sf9 His) is the recombinant Virus-derived E/Envelope protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-78506