Learn More
Medchemexpress LLC E/Envelope Dengue v 10ug
SDP
Catalog No. 5000178844
Encompass_Preferred
The NS1 protein plays a key role in viral budding association with cell membranes and facilitating the assembly of viral RNA into nucleocapsids to form mature viral particles During entry NS1 induces genome penetration into the host cytoplasm E/Envelope Protein Dengue virus 4 (395a a sf9 His) is the recombinant Virus-derived E/Envelope protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-78844 Supplier Medchemexpress LLC Supplier No. HYP7419010UG
The NS1 protein plays a key role in viral budding association with cell membranes and facilitating the assembly of viral RNA into nucleocapsids to form mature viral particles During entry NS1 induces genome penetration into the host cytoplasm E/Envelope Protein Dengue virus 4 (395a a sf9 His) is the recombinant Virus-derived E/Envelope protein expressed by Sf9 insect cells with C-His labeled tag