Learn More
Medchemexpress LLC (E)-ethyl cinnamate | 4192-77-2 | MFCD00009189 | 98.5% | 176.21 | C11H12O2 | 250 G
SDP
Catalog No. 5000283214
Encompass_Preferred
(E)-Ethyl cinnamate is the trans (E) isomer of ethyl cinnamate, an ester of cinnamic acid used as a research reagent and building block in chemical synthesis, pharmacology, and analytical studies. It is supplied at high purity for laboratory applications and accompanied by technical documents for safe handling and quality control.
  • High purity: 98.5%.
  • Molecular weight 176.21 and formula C11H12O2.
  • Available as a 250 G pack for laboratory use.
  • Stable at room temperature with a three-year shelf life; solutions should be stored at -20°C (one year) or -80°C (two years).
  • Includes datasheet, certificate of analysis, and safety data sheet for compliance.
Catalog No. 50-002-83214 Supplier Medchemexpress LLC Supplier No. HYW236273250G
(E)-Ethyl cinnamate is the trans (E) isomer of ethyl cinnamate, an ester of cinnamic acid used as a research reagent and building block in chemical synthesis, pharmacology, and analytical studies. It is supplied at high purity for laboratory applications and accompanied by technical documents for safe handling and quality control.
  • High purity: 98.5%.
  • Molecular weight 176.21 and formula C11H12O2.
  • Available as a 250 G pack for laboratory use.
  • Stable at room temperature with a three-year shelf life; solutions should be stored at -20°C (one year) or -80°C (two years).
  • Includes datasheet, certificate of analysis, and safety data sheet for compliance.