Learn More
Medchemexpress LLC (E)-β-Farnesene | 18794-84-8 | 250 MG
SDP
Catalog No. 5000422013
Encompass_Preferred
(E)-β-Farnesene is an aphid alarm pheromone naturally occurring in *Phlomis aurea Decne* essential oil. This compound exhibits significant binding with the CDK2 receptor, demonstrating a value of -30.64 kcal/mol, and also shows good affinity to odorant-binding protein 3 (OBP3). Furthermore, it can act as a feeding stimulant for the sand fly *Lutzomyia longipalpis*.
  • Functions as an aphid alarm pheromone
  • Found in *Phlomis aurea Decne* essential oil
  • Exhibits good binding to the CDK2 receptor
  • Shows affinity to odorant-binding protein 3 (OBP3)
  • Acts as a feeding stimulant for sand flies
Catalog No. 50-004-22013 Supplier Medchemexpress LLC Supplier No. 250MGHYN7364
(E)-β-Farnesene is an aphid alarm pheromone naturally occurring in *Phlomis aurea Decne* essential oil. This compound exhibits significant binding with the CDK2 receptor, demonstrating a value of -30.64 kcal/mol, and also shows good affinity to odorant-binding protein 3 (OBP3). Furthermore, it can act as a feeding stimulant for the sand fly *Lutzomyia longipalpis*.
  • Functions as an aphid alarm pheromone
  • Found in *Phlomis aurea Decne* essential oil
  • Exhibits good binding to the CDK2 receptor
  • Shows affinity to odorant-binding protein 3 (OBP3)
  • Acts as a feeding stimulant for sand flies