Learn More
TARGETMOL CHEMICALS INC E-FECP-OXINDOLE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T227555MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. (E)-FeCP-oxindole is an inhibitor of VEGFR2 with IC50 of 214 nM. purity: 99%
Catalog No. 50-258-0926