Learn More
MSC Multi-Clean Disinfectant Cleaner, Hospital Grade, Concentrate, Neutral pH, 2 Liter
Catalog No. 502840041
Encompass
Multi-Clean 128 E-FECTicide Disinfectant Cleaner is a hospital-grade, neutral pH, 1:128 concentrate disinfectant designed for high-speed applications. It effectively cleans, disinfects, and deodorizes in one step, making it suitable for various professional environments. This product is EPA List N Approved for Corona Virus and kills SARS-CoV-2 in just 1 minute.
  • Hospital grade disinfectant
  • Concentrate saves money
  • Approved for spray application
  • EPA List N approved for Corona Virus
  • Kills SARS-CoV-2 in 1 minute
  • Fast kill disinfectant-cleaner
  • No-rinse, neutral pH formula
  • Cleans, disinfects, and deodorizes in one step
  • Can be used with E-Spray electrostatic sprayers
  • Dilution flexibility with multi-task container
  • Excellent storage stability (1 year minimum)
  • Freeze-thaw stable
Catalog No. 50-284-0041 Supplier MSC Supplier No. 94689668
Multi-Clean 128 E-FECTicide Disinfectant Cleaner is a hospital-grade, neutral pH, 1:128 concentrate disinfectant designed for high-speed applications. It effectively cleans, disinfects, and deodorizes in one step, making it suitable for various professional environments. This product is EPA List N Approved for Corona Virus and kills SARS-CoV-2 in just 1 minute.
  • Hospital grade disinfectant
  • Concentrate saves money
  • Approved for spray application
  • EPA List N approved for Corona Virus
  • Kills SARS-CoV-2 in 1 minute
  • Fast kill disinfectant-cleaner
  • No-rinse, neutral pH formula
  • Cleans, disinfects, and deodorizes in one step
  • Can be used with E-Spray electrostatic sprayers
  • Dilution flexibility with multi-task container
  • Excellent storage stability (1 year minimum)
  • Freeze-thaw stable